SAMPLE LOOP 2 ML CPC 1/8 1 * 1 items
Catalog # GILI21041008
Supplier: GILSON LCMS
Frequently Bought Together
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Short-term tubes for accuro®
Supplier: Dräger
VWR® pHenomenal® Electrode Stand
Supplier: VWR Collection
VWR®, Accessories for Gravimetric Dilutors
Supplier: VWR Collection
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Titration beakers
Supplier: Mettler - Toledo
VWR® pH 110/pH 110 M, Handheld pH/mV/°C Meters
Supplier: VWR Collection
Accessories for titrators
Supplier: Mettler - Toledo
VWR® PH 20 pH Meter, Handheld
Supplier: VWR Collection
Gas detection tubes for water vapour
Supplier: Dräger
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
Single gas detectors, BW Clip
Supplier: BW TECHNOLOGIES
Accessories for cell counter, EVE™
Supplier: NANOENTEK
Accessories for CytoFLEX Platform
Supplier: Beckman Coulter
Plastic cuvette for LICO
Supplier: Hach