GX-271 ASPEC SGL 4060 WITH Z DRIVE 1 * 1 items
Catalog # GILI2614007
Supplier: GILSON LCMS
Frequently Bought Together
Nessler cylinders
Supplier: witeg Labortechnik
Accessories for Titronic® 300/500 and TitroLine® 6000 / 7000
Supplier: SI Analytics
Accessories for titrators
Supplier: Mettler - Toledo
VWR® SP100 Automatic Straw Pipette, Fixed Volume
Supplier: VWR Collection
Tubes and systems for active sampling
Supplier: Dräger
VWR®, Portable Turbidity Meter
Supplier: VWR Collection
VWR® V-3000PC Visible Spectrophotometer
Supplier: VWR Collection
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
OIL IMPACTOR 0.1-1.0 MG/M3 1 * 10 items
Supplier: Dräger
Microplate readers, multi-mode, SpectraMax® M series
Supplier: Molecular Devices
Cuvettes for fluorescence measurements
Supplier: SARSTEDT
Water activity handheld sets, HP23-AW-A
Supplier: ROTRONIC INSTRUMENTS
Titrator, Titronic® 300
Supplier: SI Analytics
Conductivity meters, handheld, Seven2Go™ S3 / S7 Pro
Supplier: Mettler - Toledo
VWR®, Eluent buffers for amino acid analyzer
Supplier: VWR Chemicals
UV/Vis Calibration Standards
Supplier: HELLMA
Melting point and boiling point capillaries and tubes
Supplier: BUCHI
Plan Apochromat objective 10X/0.35 (WD=4.2mm) 1 * 1 items
Supplier: MOTIC