CHROMABOND columns PFAS BIGpack, volume: 3 mL, content of sorbent: 120 mg, material: PP, with PE filter elements, packed in 5 bags with 50 pcs., pack of 250 1 * 250 items
Catalog # MANA7300009.250
Supplier: MACHEREY-NAGEL
Frequently Bought Together