Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
2,4-DICHLORO-5-METHYLTHIENO[2,3-D]PYRIMIDINE 1 * 10 g
Catalog # PHBLPB03753-10G
Supplier:
Pharmablock (USA)
Expand image
2,4-DICHLORO-5-METHYLTHIENO[2,3-D]PYRIMIDINE 1 * 10 g
Catalog # PHBLPB03753-10G
Supplier:
Pharmablock (USA)
Frequently Bought Together
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
Cyclohexane ≥99%
Supplier: Thermo Fisher Scientific
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Methanol ≥99.8%, Ultrapure for HPLC
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
2-Propanol 99.5% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitrile, low water content ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED®, J.T.Baker®
Supplier: Avantor
Magnesium chloride 510 g/l in aqueous solution
Supplier: Hach