The water bath includes a digital temperature control and low-water sensor to prevent burn-outs.The stainless steel chamber is corrosion resistant and temperature-controlled from ambient to 99C while using the included cover. 1 * 1 items
Catalog # EDVO5939
Supplier: EDVOTEK
Frequently Bought Together