ContactUsLinkComponent
Distributors for Bottles GL 45 with 2 ports GL 14 (straight connections) , Tmax. +140°C with 2x GL 14 made of PP with PE olives + screw cap with GL45 perforated cap made of Celanex 1 * 1 items
Catalog # NEUB6095-10-451414
Distributors for Bottles GL 45 with 2 ports GL 14 (straight connections) , Tmax. +140°C with 2x GL 14 made of PP with PE olives + screw cap with GL45 perforated cap made of Celanex 1 * 1 items
Catalog # NEUB6095-10-451414
Frequently Bought Together