CFTR(inh)-172 is a selective and reversible thiazolidinone inhibitor of the cystic fibrosis transmembrane conductance regulator (CFT R Ki = 300 nM Ma et al.). CFTR is a chloride anion channel involved in the secretion of fluid in many epithelial ti 1 * 5 mg
Catalog # STMC100-0554
Supplier: STEMCELL Technologies
Frequently Bought Together