Digital Solutions
Applications
Support
About
Slovenia
Orders Link
ContactUsLinkComponent
Forceps
Forceps, straight
Forceps, straight
130 mm
USBE3032
:
USBECK Laborgeräte
Forceps, straight
USBE3032
:
USBECK Laborgeräte
Forceps, straight
130 mm
4,90 €
1 * 1 items
-
+
Made from nickel plated steel or from stainless steel.
Straight
Fine points
Length:
130 mm
Tweezers
#: 232-0088
: USBECK Laborgeräte