ContactUsLinkComponent
KJELDAHL TABLETS MISSOURI Advantages:,,Simple utilisation, universally applicable Composition:,,4,98 g K2So4 0,02 g CuSo4 * 5 H2o 1 * 1.000 Tablet
Catalog # BUCH11072629
Supplier:  BUCHI
KJELDAHL TABLETS MISSOURI Advantages:,,Simple utilisation, universally applicable Composition:,,4,98 g K2So4 0,02 g CuSo4 * 5 H2o 1 * 1.000 Tablet
Catalog # BUCH11072629
Supplier:  BUCHI
Frequently Bought Together