ContactUsLinkComponent
KJELDAHL TABLETS TITANIUM MICRO Advantages:,,Reduces the amount of chemicals required Composition:,,1,5 g K2So4, 0,045 g CuSo4 * 5 H2O, 0,045 g TiO2 1 * 1.000 items
Catalog # BUCH11072628
Supplier:  BUCHI
KJELDAHL TABLETS TITANIUM MICRO Advantages:,,Reduces the amount of chemicals required Composition:,,1,5 g K2So4, 0,045 g CuSo4 * 5 H2O, 0,045 g TiO2 1 * 1.000 items
Catalog # BUCH11072628
Supplier:  BUCHI
Frequently Bought Together