Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
[EN]COD 25 VIALS USEPA APPROVED 0-150 PP 1 * 25 Tests
CHETK-7350S
:
CHEMetrics
[EN]COD 25 VIALS USEPA APPROVED 0-150 PP 1 * 25 Tests
CHETK-7350S
:
CHEMetrics
Frequently Bought Together
Sodium hypochlorite solution 5%, J.T.Baker®
: Avantor
Urine test strips, urinalysis
: SURESCREEN
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
: VWR Chemicals
Test papers, lead acetate papers
: MACHEREY-NAGEL
VWR® Kjeldahl Tablets
: VWR Chemicals
Reagents for ammonia determination using Lovibond® MD photometers
: Lovibond
Alcometer fuel cell breathalyser
: SURESCREEN
Test kits, photometric, COD, LCK cuvette tests
: Hach
Nitrite VHR L, 500 ml
: Lovibond