ContactUsLinkComponent
Masterflex® Transfer Tubing, Puri-Sil™ Pt, 1/16" ID x 1/4" OD; 25 Ft
Masterflex® Transfer Tubing, Puri-Sil™ Pt, 1/16" ID x 1/4" OD; 25 Ft
Catalog # MFLX04691-05
Masterflex® Transfer Tubing, Puri-Sil™ Pt, 1/16" ID x 1/4" OD; 25 Ft
Catalog # MFLX04691-05
About this item
  • Environmentally Preferable:
  • Warranty:
    Manufacturer defect
  • Pk:
    25 Feet
Frequently Bought Together