ContactUsLinkComponent
POLIPROPYLENE TUBE TYPE COMECTA 30 ML, dia. 25 x 92 mm, max 64000xg 1 * 1 items
Catalog # SELE5752398
Supplier:  SELECTA
POLIPROPYLENE TUBE TYPE COMECTA 30 ML, dia. 25 x 92 mm, max 64000xg 1 * 1 items
Catalog # SELE5752398
Supplier:  SELECTA
Frequently Bought Together