Orders LinkContactUsLinkComponent
CytoFLEX SRT Software Upgrades
CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y5-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 4 active lasers (405 nm, 488 nm, 561 nm, 638 nm) and 15 fluorescence detectors in the V5-B2-Y5-R3 configuration
Catalog # BECMC65139
Supplier:   Beckman Coulter
CytoFLEX SRT Software Upgrades
Catalog # BECMC65139
Supplier:  Beckman Coulter
CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y5-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 4 active lasers (405 nm, 488 nm, 561 nm, 638 nm) and 15 fluorescence detectors in the V5-B2-Y5-R3 configuration
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Activation key to upgrade the CytoFLEX SRT V0-B2-Y5-R3 Cell Sorter with additional parameters. Upon installation the instrument will have 4 active lasers (405 nm, 488 nm, 561 nm, 638 nm) and 15 fluorescence detectors in the V5-B2-Y5-R3 configuration
  • Seq:
    3