Masterflex® Fitting, Polypropylene, Straight, Hosebarb to Thread, 3/32" ID x 10-32 UNF(M); 250/PK
Catalog # MFLX50625-01
Supplier: Avantor Fluid Handling
CE certificate
About this item
- Barb size/tubing int.Ø (port 2):3/32"
- Connection Type:Thread to Barb
- Flow pattern:Straight
- Description:Fitting, Polypropylene, Straight, Hosebarb to Thread, 3/32" ID x 10-32 UNF(M); 250/PK
- Seq:31
- Environmentally Preferable:
- Model:50625-01
- Sterilisation:Ethylene Oxide (ETO) or Autoclave
- Pk:250
- Fitting material:Polypropylene
- Thread size UNF(M) (port 2):10-32"
Frequently Bought Together