Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Reagecon GY5 Colour Reference Solution according to European Pharmacopoeia (EP) 1 * 100 mL
Catalog # REGEEPGY501
Supplier:
REAGECON
Expand image
Reagecon GY5 Colour Reference Solution according to European Pharmacopoeia (EP) 1 * 100 mL
Catalog # REGEEPGY501
Supplier:
REAGECON
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
ICP multi-element standards (28 components)
Supplier: CPACHEM
di-Sodium sulphide 0.5 mol/l in glycerine 85% 0.5 mol/l in glycerine 85% Reag. Ph. Eur. 1083901
Supplier: VWR Chemicals
VWR®, Density standards referenced at 25 °C
Supplier: VWR Chemicals
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
VWR®, High temperature viscosity standards
Supplier: VWR Chemicals
Methyl Caproate, Reference Standard
Supplier: USP
Triclocarban, Reference Standards
Supplier: USP
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
2-Butanol, Reference Standard
Supplier: USP