Masterflex® Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/Pk
Catalog # MFLX45501-66
Supplier: Avantor Fluid Handling
CE certificate
Avantor exclusive
Specifications
See all product configurations- Model:
- 45501-66
- 45508-70
- 45508-76
- 45508-66
- 45508-60
- 30800-00
- 45501-60
- Connection type (port 1):
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Female luer
- Connection type (port 2):
- 10-32 special tapered thread
- 1/4-28 UNF bottom sealing thread
- 1/4-28 UNF thread
- 10-32 UNF thread
- 1/8-27-NPT thread
- No. of ports:
- 2
- 2
- 2
- 2
- 2
- 2
- 2
- Warranty:
- 3 Months
- 3 Months
- 3 Months
- Currently Viewing
Specifications
About this item
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
- Connection Type:Female Luer to Thread
- Flow pattern:Straight
- Description:Fitting, Polycarbonate, Straight, Female Luer to Thread Adapter, 1/4-28 UNF; 25/Pk
- Seq:03
- Environmentally Preferable:
- Model:45501-66
- Connection type (port 1):Female luer
- No. of ports:2
- Warranty:3 Months
- Pk:25
- Fitting material:Polycarbonate
- Thread size UNF(M) (port 1):1/4"-28
Frequently Bought Together