PendoTech PMAT-STND Single-Use PressureMAT Pressure Monitor Benchtop Stand
Catalog # MFLX19406-95
Supplier: Avantor Fluid Handling
CE Compliant
Specifications
See all product configurations- Catalog No:
- MFLX19406-95
- MFLX19406-15
- MFLX19406-85
- MFLX19406-34
- MFLX19406-80
- MFLX19406-14
- MFLX19406-82
- MFLX19406-48
- MFLX19406-46
- MFLX19406-45
- MFLX19406-36
- MFLX19406-87
- MFLX19406-47
- MFLX19406-35
- MFLX19406-50
- MFLX19406-49
- Max. pressure:
- 76 psi
- 76 psi
- 76 psi
- Process connection:
- 1/4" x 1/4" hose barb, male luer
- 3/8" x 3/8" hose barb, male luer
- 1/2" x 1/2" hose barb, male luer
- Length:
- 304,8 cm (120")
- 182,88 cm (72")
- 365,76 cm (144")
- 213,36 cm (84")
- 365,76 cm (144")
- 182,88 cm (72")
- Warranty:
- 2 years
- 2 years
- 2 years
- 3 Months
- 1 year
- 2 years
- 1 year
- 2 years
- 2 years
- 2 years
- 3 Months
- 2 years
- 2 years
- 3 Months
- 2 years
- 2 years
- Currently Viewing
Specifications
About this item
Optimize performance of your PendoTECH® Pressure Sensor.
- Choose from a wide range of replacement parts and accessories
Replacement parts and accessories to keep your PendoTECH systems running smoothly and accurately.
- Description:Benchtop Stand for PressureMAT Monitor
- Environmentally Preferable:
- Warranty:2 years
- Pk:1
Frequently Bought Together