TUBING NORPRENE 1/16X3/16'' 1 * 500 Feet
Catalog # MFLX00278-WW
Supplier: Avantor Fluid Handling
Frequently Bought Together
Masterflex® Single-Use I/P® Gamma-Irradiated Precision Pump Tubing, Avantor®
Supplier: Avantor Fluid Handling
Tygon® F-4040-A Tubings
Supplier: Saint Gobain Life Sciences
SUCTION CUP/SYPHON SUCTIONER MADE OF SIL 1 * 10 items
Supplier: GLASWARENFABRIK KARL HECHT
5/64 X 9/64 PT TBNG 1 * 100 items
Supplier: Avantor Fluid Handling
Burkert Type 3233 316L Stainless Steel Diaphragm Valve
Supplier: Avantor Fluid Handling
Manual Tubing Pinch Valves, Delrin®
Supplier: Avantor Fluid Handling
FTNG TCXB 50.5 X 1/2 316SS 1 * 1 items
Supplier: Avantor Fluid Handling
Silicone sealing rings for GL threads without PTFE washer DURAN®
Supplier: DWK Life Sciences
PCR tubes, Multiply®
Supplier: SARSTEDT
ULTRA PLAT SIL .025X.047 50FT 1 * 1 items
Supplier: Avantor Fluid Handling
Analytical funnels, Urbanti Type
Supplier: VITLAB
Pestles
Supplier: MORGAN ADVANCED MATERIALS HALD