ContactUsLinkComponent
Module IQ/microcontroller for system 2020, for up to 20 IQ sensors, with PROFIBUS connection, automatic air pressure compensation as well as USB interface (IP 66) and Ethernet interface (IP 66) (Please order separately D-SUB connection ADA/D-SUB 9028 1 * 1 items
Catalog # WTWO471023
Supplier:  WTW ONLINE
Module IQ/microcontroller for system 2020, for up to 20 IQ sensors, with PROFIBUS connection, automatic air pressure compensation as well as USB interface (IP 66) and Ethernet interface (IP 66) (Please order separately D-SUB connection ADA/D-SUB 9028 1 * 1 items
Catalog # WTWO471023
Supplier:  WTW ONLINE
Frequently Bought Together