1-1/2 Sanitary Assy AQC STCII 1 * 5 items
Catalog # CPCCAQSSTC2331000
Supplier: CPC COLDER
Frequently Bought Together
Masterflex® Female Luer to Barb Fitting, Avantor®
Supplier: Avantor Fluid Handling
CPC® Plastic Quick-Disconnect Fittings, Complete Couplings
Supplier: Avantor Fluid Handling
Polypropylene Fitting Manifolds, Female NPT
Supplier: Avantor Fluid Handling
3/4'' F X 1/2'' M FLARE REDUCER 1 * 1 items
Supplier: Avantor Fluid Handling
Masterflex® Cleanroom 1" Sanitary Flange to Hose Barb, Avantor®
Supplier: Avantor Fluid Handling
Value Plastics® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene
Supplier: Avantor Fluid Handling
Adapter 1/2 Tc X 1/8 Barb 1.250 Oal 316L No.7 1 * 1 items
Supplier: Avantor Fluid Handling
CPC® Plastic Quick-Disconnect Fittings, Hose Barb Inserts
Supplier: Avantor Fluid Handling
Masterflex® Reducer Fittings, Hose Barb, Straight, Stainless Steel
Supplier: Avantor Fluid Handling
CPC® High-Flow Quick-Disconnect Fittings, Threaded Bodies
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, HDPE, Avantor®
Supplier: Avantor Fluid Handling
HFC30039M SEALING PLUG HFC39 10PK 1 * 10 items
Supplier: Avantor Fluid Handling
MPC series ¹/₄" flow, quick disconnect coupling for bio and pharma
Supplier: CPC COLDER
O-RING 1 * 100 items
Supplier: Avantor Fluid Handling
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, HDPE, Avantor®
Supplier: Avantor Fluid Handling
Masterflex® Barbed High Flow Female Quick-Disconnect Fittings, Avantor®
Supplier: Avantor Fluid Handling