ContactUsLinkComponent
SONOREX DIGITEC DT 255 H-RC, ultrasonic bath, 230 V(± 10%) 50/60 Hz, ID 300x150x150 mm 5,5 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15 ,30 min and 1 * 1 items
Catalog # BAND3081
SONOREX DIGITEC DT 255 H-RC, ultrasonic bath, 230 V(± 10%) 50/60 Hz, ID 300x150x150 mm 5,5 l, s/s digital display timer 1, 2, 3, 4, 5, 10, 15 ,30 min and 1 * 1 items
Catalog # BAND3081
Frequently Bought Together