Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
20 ML MICROSTANDARD 5 ΜMa 1 * 20 mL
Catalog # MSPP11050-20
Supplier:
MARKET SOURCE PART PROCESS
Expand image
20 ML MICROSTANDARD 5 ΜMa 1 * 20 mL
Catalog # MSPP11050-20
Supplier:
MARKET SOURCE PART PROCESS
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach