ContactUsLinkComponent
Hooded coverall combining Tyvek with a light SMS back panel from shoulders to ankle. Elasticated wrists, waist and ankles. Zipper flap. Thumbhole on sleeve. Bulk packed, no individual garment packaging reducing the waste. White. Antistatic. Size XL 1 * 25 items
Catalog # DUPOTD0127XLWH00
Supplier:  DuPont
undefined
Hooded coverall combining Tyvek with a light SMS back panel from shoulders to ankle. Elasticated wrists, waist and ankles. Zipper flap. Thumbhole on sleeve. Bulk packed, no individual garment packaging reducing the waste. White. Antistatic. Size XL 1 * 25 items
Catalog # DUPOTD0127XLWH00
Supplier:  DuPont