ContactUsLinkComponent
Hooded coverall combining Tyvek with a light SMS back panel from shoulders to ankle. Elasticated wrists, waist and ankles. Zipper flap. Thumbhole on sleeve. Bulk packed, no individual garment packaging reducing the waste. White. Antistatic. Size 4XL 1 * 25 items
Catalog # DUPOTD01274XLWH00
Supplier:  DuPont
undefined
Hooded coverall combining Tyvek with a light SMS back panel from shoulders to ankle. Elasticated wrists, waist and ankles. Zipper flap. Thumbhole on sleeve. Bulk packed, no individual garment packaging reducing the waste. White. Antistatic. Size 4XL 1 * 25 items
Catalog # DUPOTD01274XLWH00
Supplier:  DuPont