Orders LinkContactUsLinkComponent
Cryogenic markers, SCIENCE-MARKER™
Cryogenic markers, SCIENCE-MARKER™
Cryogenic permanent marker, Shipping weight 0,113 kg, 15,9×101,6×168,3 mm, Black
Catalog # GAINSM-1B-6
Supplier:   GA
Cryogenic markers, SCIENCE-MARKER™
Catalog # GAINSM-1B-6
Supplier:  GA
Cryogenic markers, SCIENCE-MARKER™
Cryogenic markers, SCIENCE-MARKER™
Cryogenic permanent marker, Shipping weight 0,113 kg, 15,9×101,6×168,3 mm, Black
Price
28,80 €
Log in for availability
1 * 6 items
Product Details & Documents
Cryogenic permanent marker with fine tip.
  • Alcohol and water resistant
  • Compatible with Lab-TAG™ labels
Specifications
  • Colour:
    Black
  • Description:
    Cryogenic permanent marker, Shipping weight 0,113 kg, 15,9×101,6×168,3 mm
  • Seq:
    01