ContactUsLinkComponent
0.6% Gel Cups (TAE) w/ GelGreen DNA Stain: includes: Ten GelCups with preweighed 0.6% Agarose and GelGreen DNA Stain in 0.5XTAEbuffer, 100 mL Tris-Acetate-EDTA (TAE) buffer concentrate 1 * 10 items
Catalog # EMBIM3241TAE
Supplier:  EMBITEC
0.6% Gel Cups (TAE) w/ GelGreen DNA Stain: includes: Ten GelCups with preweighed 0.6% Agarose and GelGreen DNA Stain in 0.5XTAEbuffer, 100 mL Tris-Acetate-EDTA (TAE) buffer concentrate 1 * 10 items
Catalog # EMBIM3241TAE
Supplier:  EMBITEC
Frequently Bought Together