Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
BioMarin Validation Kit 4 - Consists of: (120) of COVMGTVKUB16X26SS (30) of BBMGTVKSS1626 (300) of COVMGTVKUB4X16SS (250) of BBMGTVKSS0415 1 * 1 KIT
129-2824
:
KEYSTONE CLEANROOM PRODUCTS
BioMarin Validation Kit 4 - Consists of: (120) of COVMGTVKUB16X26SS (30) of BBMGTVKSS1626 (300) of COVMGTVKUB4X16SS (250) of BBMGTVKSS0415 1 * 1 KIT
129-2824
:
KEYSTONE CLEANROOM PRODUCTS
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Facial Tissues
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals