ContactUsLinkComponent
Biotinylated Rat CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag (CDG-R82D4) is expressed from human 293 cells (HEK293). It conta ins AA Gln 21 - Asp 103 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_001101610.1 (CD3E) & NP_001071114.1 (CD3G)). 1 * 25 µG
Catalog # ACRBCDG-R82D4-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Rat CD3E&CD3G Heterodimer Protein, His,Avitag&Flag Tag (CDG-R82D4) is expressed from human 293 cells (HEK293). It conta ins AA Gln 21 - Asp 103 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_001101610.1 (CD3E) & NP_001071114.1 (CD3G)). 1 * 25 µG
Catalog # ACRBCDG-R82D4-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together