ContactUsLinkComponent
Biotinylated Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgG1 (AM130) is a chimeric monoclonal antibody recombinantly ex pressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 constant 1 * 25 µG
Catalog # ACRBS1N-M13L3-25UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Anti-SARS-CoV-2 Spike RBD Antibody, Chimeric mAb, Human IgG1 (AM130) is a chimeric monoclonal antibody recombinantly ex pressed from HEK293 cells, which combines the variable region of a mouse monoclonal antibody with human IgG1 constant 1 * 25 µG
Catalog # ACRBS1N-M13L3-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together