Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (SPD-C82Ec) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452R, E484Q)). The L452R, E484Q mutations were identified in the SARS-CoV- 1 * 200 µG
Catalog # ACRBSPD-C82EC200UG
Supplier: ACRO Biosystems
Frequently Bought Together