Cynomolgus NKp46, Llama IgG2b Fc Tag (NC1-C5255) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Access ion # Q95JB9-1). 1 * 1 mg
Cynomolgus NKp46, Llama IgG2b Fc Tag (NC1-C5255) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Access ion # Q95JB9-1). 1 * 1 mg