ContactUsLinkComponent
Cynomolgus NKp46, Llama IgG2b Fc Tag (NC1-C5255) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Access ion # Q95JB9-1). 1 * 1 mg
Catalog # ACRBNC1-C5255-1MG
Supplier:  ACRO Biosystems
Cynomolgus NKp46, Llama IgG2b Fc Tag (NC1-C5255) is expressed from human 293 cells (HEK293). It contains AA Pro 22 - Arg 257 (Access ion # Q95JB9-1). 1 * 1 mg
Catalog # ACRBNC1-C5255-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together