ContactUsLinkComponent
Human Tissue Factor, His Tag (TF3-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 251 (Accession # P1 3726-1). 1 * 500 µG
Catalog # ACRBTF3-H52H5500UG
Supplier:  ACRO Biosystems
Human Tissue Factor, His Tag (TF3-H52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 33 - Glu 251 (Accession # P1 3726-1). 1 * 500 µG
Catalog # ACRBTF3-H52H5500UG
Supplier:  ACRO Biosystems
Frequently Bought Together