ContactUsLinkComponent
Mouse Latent TGFB1, His Tag (TG1-M5245) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P04 202-1). 1 * 1 mg
Catalog # ACRBTG1-M5245-1MG
Supplier:  ACRO Biosystems
Mouse Latent TGFB1, His Tag (TG1-M5245) is expressed from human 293 cells (HEK293). It contains AA Leu 30 - Ser 390 (Accession # P04 202-1). 1 * 1 mg
Catalog # ACRBTG1-M5245-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together