ContactUsLinkComponent
Mouse Tissue Factor, His Tag (TF3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Glu 251 (Accession # P2 0352-1). 1 * 500 µG
Catalog # ACRBTF3-M52H3500UG
Supplier:  ACRO Biosystems
Mouse Tissue Factor, His Tag (TF3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 29 - Glu 251 (Accession # P2 0352-1). 1 * 500 µG
Catalog # ACRBTF3-M52H3500UG
Supplier:  ACRO Biosystems
Frequently Bought Together