Human FGL1 (64-312), Fc Tag (FG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08 830-1). 1 * 1 mg
ACRBFG1-H5254-1MG
Human FGL1 (64-312), Fc Tag (FG1-H5254) is expressed from human 293 cells (HEK293). It contains AA Asp 64 - Ile 312 (Accession # Q08 830-1). 1 * 1 mg
ACRBFG1-H5254-1MG
Frequently Bought Together