ContactUsLinkComponent
Human Apolipoprotein E (R154S,R176C), His Tag (APE-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 31 7 (R154S,R176C) (Accession # P02649-1 (R154S,R176C)). 1 * 100 µG
Catalog # ACRBAPE-H5256100UG
Supplier:  ACRO Biosystems
Human Apolipoprotein E (R154S,R176C), His Tag (APE-H5256) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 31 7 (R154S,R176C) (Accession # P02649-1 (R154S,R176C)). 1 * 100 µG
Catalog # ACRBAPE-H5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together