Human Carbonic Anhydrase IX / CA9 (38-414), Fc Tag (CA9-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - A sp 414 (Accession # Q16790-1). 1 * 50 µG
Human Carbonic Anhydrase IX / CA9 (38-414), Fc Tag (CA9-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 38 - A sp 414 (Accession # Q16790-1). 1 * 50 µG