Mouse Carbonic Anhydrase IX / CA9 (32-390), His Tag (CA9-M52H3) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Asp 390 (Accession # Q8VHB5-1). 1 * 50 µG
Catalog # ACRBCA9-M52H3-50UG
Supplier: ACRO Biosystems
Frequently Bought Together