Rat 4-1BB Ligand His,Flag Tag (41L-R52D3) is expressed from human 293 cells (HEK293). It contains AA Pro 106 - Gly 308 (Accession # Q80WE6-1). 1 * 100 µG
Rat 4-1BB Ligand His,Flag Tag (41L-R52D3) is expressed from human 293 cells (HEK293). It contains AA Pro 106 - Gly 308 (Accession # Q80WE6-1). 1 * 100 µG