ContactUsLinkComponent
PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is produced via site-specific conjugation of PE to Human Glypican 3 (25-559), His T ag under optimal conditions with a proprietary technology.PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is expres 1 * 200 Tests
Catalog # ACRBGP3-HP2E3-200
Supplier:  ACRO Biosystems
PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is produced via site-specific conjugation of PE to Human Glypican 3 (25-559), His T ag under optimal conditions with a proprietary technology.PE-Labeled Human Glypican 3, His Tag (GP3-HP2E3) is expres 1 * 200 Tests
Catalog # ACRBGP3-HP2E3-200
Supplier:  ACRO Biosystems
Frequently Bought Together