ContactUsLinkComponent
PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (25-378), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is expressed from 1 * 200 Tests
Catalog # ACRBEGI-HP2E3-200
Supplier:  ACRO Biosystems
PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is produced via site-specific conjugation of PE to Human EGFRvIII (25-378), His Tag under optimal conditions with a proprietary technology.PE-Labeled Human EGFRvIII, His Tag (EGI-HP2E3) is expressed from 1 * 200 Tests
Catalog # ACRBEGI-HP2E3-200
Supplier:  ACRO Biosystems
Frequently Bought Together