Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1). 1 * 100 µG
Catalog # ACRBTLA-H5243100UG
Supplier: ACRO Biosystems
Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1). 1 * 100 µG
Catalog # ACRBTLA-H5243100UG
Supplier: ACRO Biosystems
Frequently Bought Together
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Amersham™ Protran® Western Blotting Membranes, Nitrocellulose
Supplier: Cytiva
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Petri dishes
Supplier: Greiner Bio-One
Chromogenic endotoxin quant kit, Pierce™
Supplier: Thermo Fisher Scientific