ContactUsLinkComponent
Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1). 1 * 100 µG
Catalog # ACRBTLA-H5243100UG
Supplier:  ACRO Biosystems
Human TL1A, His Tag (TLA-H5243) is expressed from human 293 cells (HEK293). It contains AA Leu 72- Leu 251 (Accession # O95150-1). 1 * 100 µG
Catalog # ACRBTLA-H5243100UG
Supplier:  ACRO Biosystems
Frequently Bought Together