ContactUsLinkComponent
Canine IL-2 R alpha, His Tag (ILA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 237 (Accession # O6 2802-1). 1 * 100 µG
Catalog # ACRBILA-C52H4100UG
Supplier:  ACRO Biosystems
Canine IL-2 R alpha, His Tag (ILA-C52H4) is expressed from human 293 cells (HEK293). It contains AA Asp 22 - Gln 237 (Accession # O6 2802-1). 1 * 100 µG
Catalog # ACRBILA-C52H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together