ContactUsLinkComponent
Rat LIV-1, His Tag (LV1-R52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Trp 311 (Accession # Q4V887-1). 1 * 100 µG
Catalog # ACRBLV1-R52H9100UG
Supplier:  ACRO Biosystems
Rat LIV-1, His Tag (LV1-R52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Trp 311 (Accession # Q4V887-1). 1 * 100 µG
Catalog # ACRBLV1-R52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together