Biotinylated Mouse PSCA, His,Avitag (PSA-M82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Asn 95 (Accessio n # P57096-1). 1 * 25 µG
Biotinylated Mouse PSCA, His,Avitag (PSA-M82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Asn 95 (Accessio n # P57096-1). 1 * 25 µG