Rat Nectin-4, Fc Tag (NE4-R5255) is expressed from human 293 cells (HEK293). It contains AA Tyr 28 - Val 348 (Accession # D3ZET1-1). 1 * 100 µG
ACRBNE4-R5255100UG
Rat Nectin-4, Fc Tag (NE4-R5255) is expressed from human 293 cells (HEK293). It contains AA Tyr 28 - Val 348 (Accession # D3ZET1-1). 1 * 100 µG
ACRBNE4-R5255100UG
Frequently Bought Together