ContactUsLinkComponent
Human Semaphorin 3A, Fc Tag (SEA-H5257) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Val 771 (Accession # Q14 563-1 (R552A, R555A)). 1 * 100 µG
Catalog # ACRBSEA-H5257100UG
Supplier:  ACRO Biosystems
Human Semaphorin 3A, Fc Tag (SEA-H5257) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Val 771 (Accession # Q14 563-1 (R552A, R555A)). 1 * 100 µG
Catalog # ACRBSEA-H5257100UG
Supplier:  ACRO Biosystems
Frequently Bought Together