ContactUsLinkComponent
Cynomolgus ITGA7X1B&ITGB1 Heterodimer Protein, Tag Free&His Tag (IT1-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Pro 1041 (ITGA7X1B) & Gln 21 - Asp 728 (ITGB1) (Accession # XP_005571182.1 (ITGA7X1B) & A0A2K5WHU8-1 (ITGB 1 * 100 µG
Catalog # ACRBIT1-C52W3100UG
Supplier:  ACRO Biosystems
Cynomolgus ITGA7X1B&ITGB1 Heterodimer Protein, Tag Free&His Tag (IT1-C52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Pro 1041 (ITGA7X1B) & Gln 21 - Asp 728 (ITGB1) (Accession # XP_005571182.1 (ITGA7X1B) & A0A2K5WHU8-1 (ITGB 1 * 100 µG
Catalog # ACRBIT1-C52W3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together