ContactUsLinkComponent
Mouse TPBG, His Tag (TPG-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 361 (Accession # Q9Z0L0-1). 1 * 100 µG
Catalog # ACRBTPG-M52H3100UG
Supplier:  ACRO Biosystems
Mouse TPBG, His Tag (TPG-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 361 (Accession # Q9Z0L0-1). 1 * 100 µG
Catalog # ACRBTPG-M52H3100UG
Supplier:  ACRO Biosystems
Frequently Bought Together